Magnetic stirrer, MIX 1 XL
Supplier: 2MAG
30201_UK 30201_CH 30201
442-0805EA 988 EUR
442-0805 442-0804 442-0803
Magnetic stirrer, MIX 1 XL
Stirrers Magnetic Stirrers
Robust magnetic stirrer with hermetically-sealed stainless steel housing, suitable for highly viscous media. Adjustable interval times for automated restart after a maximum of 240 s. Operating temperatures up to 50 °C. Power setting in four steps.
  • Very strong magnetic coupling via SmCo magnets, for magnetic power transmission over extended distances and thick vessel bottoms
  • Adjustable SoftStart function for reliable and safe stirring bar acceleration (20/30/60/120 seconds)
  • QuickSet for setting of start and maximum speed
  • QuickStop function against after-run and glass breakage
  • Storage of last working status when turning off
  • Digital display for speed, power, SoftStart and interval settings
Equipped as standard with mixWATCH ensuring very high process safety, e.g. when using closed non glass vessels, or for unattended overnight operation. In case of the magnetic coupling’s separation between the magnetic stirrer and the stirring bar during the acceleration phase, the magnetic stirrer will stop, the stirring bar centred and stirring resumed with SoftStart.
IP protection class: IP 64
Certifications: Protection class: IP 64