1,10-Phenanthroline hydrochloride monohydrate, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: 1,10-Phenanthroline Chloride Monohydrate, 1,10-Phenanthrolinium chloride monohydrate, 1,10-Phenanthroline monohydrochloride monohydrate
M-Clarity™ quality level = MQ100
P13002-10G P13002-25G
SIALP13002-10GEA 76.7 EUR
SIALP13002-10G SIALP13002-25G
1,10-Phenanthroline hydrochloride monohydrate, Sigma-Aldrich®
1,10-Phenanthroline hydrochloride monohydrate
Formula: C₁₂H₉ClN₂·1H₂O
MW: 234.68 g/mol
Melting Pt: 224…226 °C
Density: 1.385 g/cm³ (–100.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150061
CAS Number: 18851-33-7
EINECS: 223-325-1
UN: 2811
ADR: 6.1 III