1,2,4,5-Tetracyanobenzene, Sigma-Aldrich®

Supplier: Merck

391980-1G 391980-5G
SIAL391980-1GEA 34.4 EUR
SIAL391980-1G SIAL391980-5G
1,2,4,5-Tetracyanobenzene, Sigma-Aldrich®
1,2,4,5-Tetracyanobenzene
Formula: C₁₀H₂N₄
Boiling Pt: 374 °C (1013 hPa)
Melting Pt: 272 °C
Density: 1.372 g/cm³ (20 °C)
MDL Number: MFCD00043481
CAS Number: 712-74-3
UN: 3439
ADR: 6.1,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR