Methyl-2-bromo-5-nitrobenzoate ≥98%
Supplier: Thermo Fisher Scientific
|
Warning
|
B21288.06
B21288.14
B21288.06EA
60.3
EUR
B21288.06
B21288.14
Methyl-2-bromo-5-nitrobenzoate ≥98%
Methyl 2-bromo-5-nitrobenzoate
Purity: 98+%
CAS No.: 6942-36-5
Molecular Formula: C8H6BrNO4
Formula Weight: 260.05
Melting Point: 79-82°
Beilstein Reference: 2617575
Formula:
C₈H₆BrNO₄ Melting Pt: 79…82 °C Storage Temperature: Ambient |
MDL Number:
MFCD00010867 CAS Number: 6942-36-5 |