Thiamine hydrochloride, Sigma-Aldrich®
Supplier: Merck
Synonyms: Aneurine hydrochloride, 3-((4-Amino-2-methylpyrimidin-5-yl)methyl)-5-(2-hydroxyethyl)-4-methylthiazol-3-ium chloride hydrochloride, Thiaminium chloride, VitamineB1 hydrochloride
M-Clarity™ quality level = MQ300
W332208-1KG-K W332208-5KG-K W332208-10KG-K
SIALW332208-1KG-KEA 193 EUR
SIALW332208-1KG-K SIALW332208-5KG-K SIALW332208-10KG-K
Thiamine hydrochloride, Sigma-Aldrich®
Thiamine hydrochloride
Formula: C₁₂H₁₈Cl₂N₄OS
MW: 337.27 g/mol
Melting Pt: 252
Density: 1.401 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00012780
CAS Number: 67-03-8
EINECS: 200-641-8
Merck Index: 13,09366