Diisobutylaluminium hydride 1.1 M cyclohexane, AcroSeal®
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Di-i-butylaluminum hydride, DIBAL-H
201031000 201038000
201031000.EA 81.1 EUR
201031000. 201038000.
Diisobutylaluminium hydride 1.1 M cyclohexane, AcroSeal®
Diisobutylaluminium hydride
Formula: C₈H₁₉Al
MW: 142.22 g/mol
Melting Pt: –70 °C
Density: 0.798
Flash Pt: –018 °C
Storage Temperature: Ambient
MDL Number: MFCD00008928
CAS Number: 1191-15-7
EINECS: 214-729-9
UN: 3399
ADR: 4.3 I
Order Now
Specification Test Results
Appearance Clear solution
Strength 19.5 to 20.5 % (DIBAL-H, on Al basis)