Anti-YAP1 Recombinant Recombinant Antibody [clone: DMC495]
Supplier: ANTIBODIES.COM
A318698-100 A318698-500
ANTIA318698-100EA 640 EUR
ANTIA318698-100 ANTIA318698-500
Anti-YAP1 Recombinant Recombinant Antibody [clone: DMC495]
Antibodies
Recombinant chimeric monoclonal [DMC495] antibody to YAP1 for flow cytometry with samples derived from human.
  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 1:100
Type: Primary
Antigen: YAP1
Clonality: Recombinant
Clone: DMC495
Conjugation: Unconjugated
Epitope:
Host: Recombinant
Isotype: IgG1
Reactivity: Human