Anti-GFAP Mouse Monoclonal Antibody [Clone: 3E10]

Supplier: ANTIBODIES.COM
A270551-100 A270551-50
ANTIA270551-100EA 520 EUR
ANTIA270551-100 ANTIA270551-50
Anti-GFAP Mouse Monoclonal Antibody [Clone: 3E10]
Antibodies
Mouse monoclonal [3E10] antibody to GFAP for WB, ICC/IF and IHC with samples derived from Human, Rat, Mouse, Porcine and Bovine.

  • Validated applications: WB, ICC/IF, IHC
  • Recommended dilutions: WB: 1:1000, ICC/IF: 1:500, IHC: 1:500

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 3E10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Mouse, Porcine, Bovine
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR