Anti-IgG Goat Polyclonal Antibody (mFluor™ Violet 510)

Supplier: AAT BIOQUEST
49250 49251
AATB49250EA 198 EUR
AATB49250 AATB49251
Anti-IgG Goat Polyclonal Antibody (mFluor™ Violet 510)
Antibodies
mFluor™ Violet 510 goat anti-rabbit conjugates are secondary antibodies designed for optimal performance in immunoassay applications, including flow cytometry, immunofluorescence, and confocal microscopy.

  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography

mFluor™ Violet 510 is optimally excited by the violet laser and emits bright green fluorescence maximally at 505 nm.

Caution: Research use only.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: mFluor™ Violet 510
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR