Anti-IgG Goat Polyclonal Antibody (mFluor™ Violet 510)
Supplier: AAT BIOQUEST
49570 49571
AATB49570EA 266 EUR
AATB49570 AATB49571
Anti-IgG Goat Polyclonal Antibody (mFluor™ Violet 510)
Antibodies
mFluor™ Violet 510 goat anti-rabbit conjugates are secondary antibodies designed for optimal performance in immunoassay applications, including flow cytometry, immunofluorescence, and confocal microscopy.
  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography
mFluor™ Violet 510 is optimally excited by the violet laser and emits bright green fluorescence maximally at 505 nm.
Caution: Research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: mFluor™ Violet 510
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Rabbit