CHROMAFIL® Syringe Filters
Supplier: MACHEREY-NAGEL
729021 729022 729030 729023 729004 729020 729009 729007 729006 729008 729010 729011 729012 729013 729014 729015 729024 729025 729026 729027 729028 729031 729032 729033 729004.400 729007.400 729012.400 729013.400 729020.400 729021.400 729026.400 729027.400 729028.400 729030.400 729031.400 729032.400 729033.400 729034 729205 729219 729205.400 729221.400 729231.400 729228.400 729022.800 729218 729229 729037 729036 729241 729023.800 729242 729221 729241.400 729206 729212 729212.400 729207.400 729220 729220.400 729228 729229.400 729204 729204.400 729240 729226 729226.400 729227 729230 729231 729213 729213.400 729219.400 729218.400 729051 729207 729039 729227.400 729246 729254 729257 729230.400 729236 729237 729050 729050.400 729051.400 729039.400 729243 729244 729245 729246.400 729256 729247 729043 729209 729208 729222 729223 729248 729249 729258 729048 729044 729049 729234 729255 729260 729261 729270 729271
516-6030EA 129 EUR
516-6030 MANA729022 MANA729030 516-0386 MANA729004 516-0384 MANA729009 MANA729007 MANA729006 MANA729008 MANA729010 MANA729011 MANA729012 MANA729013 MANA729014 MANA729015 MANA729024 MANA729025 MANA729026 MANA729027 MANA729028 516-0387 MANA729032 MANA729033 MANA729004.400 MANA729007.400 MANA729012.400 MANA729013.400 516-0385 MANA729021.400 MANA729026.400 MANA729027.400 MANA729028.400 MANA729030.400 MANA729031.400 MANA729032.400 516-0388 MANA729034 514-0780 514-0723 MANA729205.400 MANA729221.400 514-0707 MANA729228.400 MANA729022.800 514-0722 MANA729229 MANA729037 MANA729036 MANA729241 MANA729023.800 MANA729242 514-0777 MANA729241.400 514-0718 514-0784 MANA729212.400 MANA729207.400 514-0778 MANA729220.400 MANA729228 MANA729229.400 514-0719 514-0721 MANA729240 514-0782 MANA729226.400 514-0783 514-0702 514-0703 514-0785 MANA729213.400 514-0727 514-0726 514-0709 514-0779 514-0728 MANA729227.400 514-0713 514-0781 514-0715 514-0706 514-0704 514-0705 514-0708 514-0710 514-0711 514-0729 514-0724 514-0725 514-0712 514-0717 514-0714 MANA729247 MANA729043 MANA729209 MANA729208 MANA729222 MANA729223 MANA729248 MANA729249 MANA729258 MANA729048 MANA729044 MANA729049 MANA729234 MANA729255 MANA729260 MANA729261 MANA729270 MANA729271
CHROMAFIL® Syringe Filters
Filters Syringe Filters
These syringe filters are used for filtration of suspended matter from liquid samples.
By using CHROMAFIL® syringe filters with their specific membranes unwanted particles are removed which can otherwise cause contamination and clogging of sensitive instrumentation.
CHROMAFIL® Xtra (labelled)
• Broad range of different membrane types
• Imprint for direct identification of membrane type, diameter and pore size
• Low-bleeding PP housing
• Colour free clear PP
CHROMAFIL® (colour coded)
• Different membrane types
• Coloured PP housing for identification of membrane type
CHROMAFIL® Combi
• Syringe filters with integrated glass fiber prefilter
• For solutions with a high load of particulate matter
PET - Hydrophilic multipurpose polyester membrane for polar as well as nonpolar solvents; especially suited for mixtures of water and organic solvents (HPLC eluents).
RC - Hydrophilic regenerated cellulose membrane for aqueous and organic-aqueous liquids, i.e. polar and medium polar sample solutions.
PTFE - Hydrophobic polytetrafluoroethylene membrane for nonpolar liquids and gases; very resistant towards all kinds of solvents as well as acids and bases.
H-PTFE - Hydrophilised polytetrafluoroethylene membrane (hydrophobic with additional hydrophilic characteristic) for polar and nonpolar solutions; resistant towards all kinds of solvents as well as acids and bases.
MV - Hydrophilic cellulose mixed esters membrane with very low adsorption for aqueous or polar solutions.
CA - Hydrophilic cellulose acetate membrane for filtration of water-soluble oligomers and polymers; especially suited for biological macromolecules; very high stability in aqueous solutions.
PA - Moderately hydrophilic polyamide (Nylon) membrane for aqueous and organic-aqueous medium polar liquids.
PES - Hydrophilic polyethersulfone membrane for aqueous and slightly organic liquids; very low adsorption for pharmaceuticals and proteins; good stability against acids and bases.
PVDF - Hydrophilic polyvinylidene difluoride membrane for 100% aqueous samples; water-soluble oligomers and polymers like proteins.
GF - Inert glass fiber filter with nominal pore size of 1 µm allows higher flow rates than small pore filters; for solutions with high loads of particulate matter or for highly viscous solutions (e.g., soil samples, fermentation broths). Also available as integrated prefilter combined with a specific membrane.
IC - Special filter for the filtration of aqueous liquids in ion chromatography. (For optimal results with blind values <5 ppb a pre-washing with de-ionized water is recommended).
All filters can be autoclaved at 121 °C and 1,1 bar for 30 min.
Inlet/outlet: Luer lock female/Luer male.
MS = minispike outlet