3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) ≥98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
B21927.03 B21927.06 B21927.14
B21927.03EA 56.3 EUR
B21927.03 B21927.06 B21927.14
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS) ≥98%
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Purity: 98+% CAS No.: 75621-03-3 Molecular Formula: C32H58N2O7S Formula Weight: 614.88 Melting Point: 156-158° dec. MDL No.: MFCD00012116
Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Melting Pt: 156…158 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00012116
CAS Number: 75621-03-3
Merck Index: 13,02052