3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS), Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
M-Clarity™ quality level = MQ400
C5070-1G C5070-5G
SIALC5070-1GEA 131 EUR
SIALC5070-1G SIALC5070-5G
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS), Sigma-Aldrich®
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Melting Pt: 116
Storage Temperature: Refrigerator
MDL Number: MFCD00081083
CAS Number: 75621-03-3
Merck Index: 13,02052