3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS), Sigma-Aldrich®
Supplier: Merck
Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS
M-Clarity™ quality level = MQ400
C3023-100G C3023-1G C3023-25G C3023-5G
SIALC3023-100GEA 2940 EUR
SIALC3023-100G SIALC3023-1G SIALC3023-25G SIALC3023-5G
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS), Sigma-Aldrich®
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Melting Pt: 116
Storage Temperature: Refrigerator
MDL Number: MFCD00081083
CAS Number: 75621-03-3
Merck Index: 13,02052