3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Supplier: PanReac AppliChem
A1099.0005
A1099.0050
A1099.0005EA
99.9
EUR
A1099.0005
A1099.0050
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula:
C₃₂H₅₈N₂O₇S Melting Pt: 116 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00081083 CAS Number: 75621-03-3 Merck Index: 13,02052 |