(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate), Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: (3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxypropanesulphonate), CHAPSO
M-Clarity™ quality level = MQ200
C3649-1G C3649-25G C3649-500MG C3649-5G
SIALC3649-1GEA 205 EUR
SIALC3649-1G SIALC3649-25G SIALC3649-500MG SIALC3649-5G
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate), Sigma-Aldrich®
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Formula: C₃₂H₅₈N₂O₈S
MW: 630.89 g/mol
Melting Pt: 184…186 °C
Storage Temperature: Refrigerator
CAS Number: 82473-24-3