(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Supplier: BIOMOL RESEARCH LABORATORIES
Warning
50792-1 50792-2
BMOL50792-1EA 0 EUR
BMOL50792-1 BMOL50792-2
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
(3-((3-Cholamidopropyl)dimethylammonio)-2-hydroxy-1-propanesulphonate)
Formula: C₃₂H₅₈N₂O₈S
Melting Pt: 184…186 °C
Storage Temperature: Refrigerator
CAS Number: 82473-24-3