Chromotropic acid disodium salt dihydrate, Sigma-Aldrich®
Supplier: Merck
|
Warning
|
Synonyms:
4,5-Dihydroxynaphthalene-2,7-disulphonic acid disodium salt dihydrate, Disodium 4,5-dihydroxynaphthalene-2,7-disulphonate dihydrate, 4,5-Dihydroxynaphthalene-2,7-disulphonic acid, disodium salt dihydrate, 4,5-Dihydroxy-2,7-naphthalenedisulphonic acid disodium salt dihydrate
M-Clarity™ quality level = MQ200
126225-100G
126225-500G
SIAL126225-100GEA
166
EUR
SIAL126225-100G
SIAL126225-500G
Chromotropic acid disodium salt dihydrate, Sigma-Aldrich®
Chromotropic acid disodium salt dihydrate
Formula:
(HO)₂C₁₀H₄(SO₃Na)₂·2H₂O MW: 400.29 g/mol Melting Pt: 300 Storage Temperature: Ambient |
MDL Number:
MFCD00150612 CAS Number: 5808-22-0 EINECS: 204-972-9 Merck Index: 13,02261 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...