Silica gel, granules -3/+8 mesh desiccant
Supplier: Thermo Fisher Scientific
44386.30
44386.A1
44386.A7
44386.30EA
38.5
EUR
44386.30
44386.A1
44386.A7
Silica gel, granules -3/+8 mesh desiccant
Silica gel
CAS No.: 7631-86-9
Sensitivity: Hygroscopic
Odour: Odourless
MDL No.: MFCD00011232
Formula:
SiO₂ Boiling Pt: 2230 °C (1013 hPa) Melting Pt: 1710 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011232 CAS Number: 63231-67-4 EINECS: 231-545-4 REACH: 01-2119379499-16 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...