Silica gel, Pore size: 110 Å ≥99.99%, spherical 1.6 µm APS for HPLC, UHPLC
Supplier: Thermo Fisher Scientific
Warning
45795.03 45795.06
45795.03EA 145 EUR
45795.03 45795.06
Silica gel, Pore size: 110 Å ≥99.99%, spherical 1.6 µm APS for HPLC, UHPLC
Silica gel
Purity: 99.99+% CAS No.: 63231-67-4 Physical Form: S.A. 340m2/g, P.V. 0.95cc/g MDL No.: MFCD00011232
Formula: SiO₂
Storage Temperature: Ambient
MDL Number: MFCD00011232
CAS Number: 63231-67-4
EINECS: 231-545-4