Florisil® Chromatography
Supplier: LGC Standards PROMOCHEM
Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil
SC-4179-B005 SC-4179-S020
PROOSC-4179-B005EA 277 EUR
PROOSC-4179-B005 PROOSC-4179-S020
Florisil® Chromatography
Magnesium silicate
Formula: MgO₃Si
MW: 100.38 g/mol
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1