White spirit, 150…200 °C, Sigma-Aldrich®
Supplier: Merck
Danger
M-Clarity™ quality level = MQ100
86460-1L 86460-2.5L
SIAL86460-1LEA 61.4 EUR
SIAL86460-1L SIAL86460-2.5L
White spirit, 150…200 °C, Sigma-Aldrich®
White Spirit
CAS Number: 8052-41-3
EINECS: 232-489-3
UN: 1268
ADR: 3,III