Wright's stain, pure, certified for microscopical staining
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Eosin Methylene blue solution according to Wright, Wright's eosin methylene blue
422640100 422640250 422641000
ACRO422640100EA 29.1 EUR
ACRO422640100 ACRO422640250 ACRO422641000
Wright's stain, pure, certified for microscopical staining
Wright's stain
Density: 0,8 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00082143
CAS Number: 68988-92-1
EINECS: 273-541-5
Order Now
Specification Test Results
Appearance Dark green Crystalline powder
Infrared spectrum Conforms
UV spectrum Complies
BSC Certification Passes