D-Asparagine monohydrate
Supplier: Thermo Fisher Scientific
Synonyms: (R)-2-Aminosuccinamic acid hydrate, D-2-Aminosuccinamic acid, H-D-Asn-OH·H₂O, (R)-2,4-Diamino-4-oxobutanoic acid hydrate
204270010 204270050 204270250
204270010.EA 44.7 EUR
204270010. 204270050. 204270250.
D-Asparagine monohydrate
D-Asparagine monohydrate
Formula: C₄H₁₀N₂O₄
MW: 150.14 g/mol (132.12 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00149558
CAS Number: 5794-24-1
Order Now
Specification Test Results
Appearance white crystalline powder or crystals
Infrared spectrum Conforms
Titration Total base 98.5 to 101.5 % (on dry substance)
Loss on drying 11.5 % to 12.5 % (105°C, 3 hrs)
Heavy metals (as Pb) ≤20 ppm