Boldenone

Supplier: Thermo Fisher Scientific

465751000 465755000
465751000.EA 128 EUR
465751000. 465755000.
Boldenone
Boldenone
Formula: C₁₉H₂₆O₂
MDL Number: MFCD00037712
CAS Number: 846-48-0

Order Now

Specification Test Results

Appearance white to light yellow to light beige powder or crystals
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 167°C to 172°C
Water ≤1 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR