7,4'-Dihydroxyflavanone 97%

Supplier: Thermo Fisher Scientific
Warning

469810010 469812500
469810010.EA 548 EUR
469810010. 469812500.
7,4'-Dihydroxyflavanone 97%
7,4'-Dihydroxyflavanone
Formula: C₁₅H₁₂O₄
Boiling Pt: 530°C (760mmHg)
Storage Temperature: Refrigerator
MDL Number: MFCD00287289
CAS Number: 578-86-9

Order Now

Specification Test Results

Appearance white to light beige powder or crystals
Infrared spectrum Conforms
HPLC ≥96.0 %
Water ≤5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR