Benzaldehyde-4-nitrophenylhydrazone 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: Benzal-4-nitrophenylhydrazone
470500010 470500050
470500010.EA 17.3 EUR
470500010. 470500050.
Benzaldehyde-4-nitrophenylhydrazone 98%
Benzaldehyde-4-nitrophenylhydrazone
Formula: C₁₃H₁₁N₃O₂
MW: 241.25 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00024596
CAS Number: 3078-09-9
Order Now
Specification Test Results
Appearance light yellow to brown crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Melting point 193°C to 197°C