Pirarubicin 95%
Supplier: Thermo Fisher Scientific
|
Danger
|
471110250
471111000
471110250.EA
288
EUR
471110250.
471111000.
Pirarubicin 95%
Pirarubicin
Formula:
C₂₃H₃₇NO₁₂ Boiling Pt: 834.7°C (760mmHg) Storage Temperature: Refrigerator |
MDL Number:
MFCD00869742 CAS Number: 72496-41-4 Merck Index: 13,00757 |
Specification Test Results
Appearance | orange to red-orange powder |
Infrared spectrum | Conforms |
HPLC | ≥94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...