AmberChrom™ hydrogen form 50WX8 100-200 mesh
Supplier: Thermo Fisher Scientific
Danger
472920025 472921000 472925000
472920025.EA 1420 EUR
472920025. 472921000. 472925000.
AmberChrom™ hydrogen form 50WX8 100-200 mesh
AmberChrom™ hydrogen form
Storage Temperature: Ambient
MDL Number: MFCD00132726
CAS Number: 69011-22-9
Order Now
Specification Test Results
Appearance beige to green-beige to light brown beads
Total exchange capacity ≥1.7 eq/l (H form)
Water retention capacity 50 to 58 % (H form)
Particle size 100 to 200 mesh