AmberChrom™ chloride form 1x8 50-100 mesh
Supplier: Thermo Fisher Scientific
472950025 472951000 472955000
472950025.EA 1210 EUR
472950025. 472951000. 472955000.
AmberChrom™ chloride form 1x8 50-100 mesh
AmberChrom™ chloride form
Formula: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
Storage Temperature: Ambient
MDL Number: MFCD00132718
CAS Number: 69011-19-4
Order Now
Specification Test Results
Appearance white to yellow to tan beads. Material can contain black specks per manufacturer. Does not impact use.
Total exchange capacity ≥1.2 eq/l (Cl form)
Water retention capacity 43 to 48 % (Cl form)
Particle size 50 to 100 mesh