AmberChrom™ chloride form 1x8 200-400 mesh
Supplier: Thermo Fisher Scientific
472970025 472971000 472975000
472970025.EA 1800 EUR
472970025. 472971000. 472975000.
AmberChrom™ chloride form 1x8 200-400 mesh
AmberChrom™ chloride form
Formula: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
Storage Temperature: Ambient
MDL Number: MFCD00132718
CAS Number: 69011-19-4
Order Now
Specification Test Results
Appearance white to yellow beads
Total exchange capacity ≥1.2 eq/l (Cl form)
Water retention capacity 39 to 45 % (Cl form)
Particle size 200 to 400 mesh