AmberChrom™ hydrogen form 50WX2 200-400 mesh
Supplier: Thermo Fisher Scientific
Danger
473030025 473031000 473035000
473030025.EA 1180 EUR
473030025. 473031000. 473035000.
AmberChrom™ hydrogen form 50WX2 200-400 mesh
AmberChrom™ hydrogen form
Storage Temperature: Ambient
MDL Number: MFCD00132722
CAS Number: 69011-22-9
Order Now
Specification Test Results
Appearance white to yellow to orange beads
Total exchange capacity ≥0.6 eq/l (H form)
Water retention capacity 74 to 82 % (H form)
Particle size 200 to 400 mesh