AmberChrom™ chloride form 1x2 100-200 mesh
Supplier: Thermo Fisher Scientific
473050025 473051000 473055000
473050025.EA 1350 EUR
473050025. 473051000. 473055000.
AmberChrom™ chloride form 1x2 100-200 mesh
AmberChrom™ chloride form
Formula: (C₁₀H₁₂·C₁₀H₁₀·C₈H₈·C₃H₉N)x
Storage Temperature: Ambient
MDL Number: MFCD00146763
CAS Number: 69011-19-4
Order Now
Specification Test Results
Appearance white to yellow beads
Total exchange capacity ≥0.6 eq/l (Cl form)
Water retention capacity 65 to 75 % (Cl form)
Particle size 100 to 200 mesh