4-Aminohippuric acid, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ200

A1422-100G A1422-10G A1422-25G
SIALA1422-100GEA 532 EUR
SIALA1422-100G SIALA1422-10G SIALA1422-25G
4-Aminohippuric acid, Sigma-Aldrich®
4-Aminohippuric acid
Formula: H₂NC₆H₄C(O)NHCH₂COOH
Boiling Pt: 441 °C (1013 hPa)
Melting Pt: 199…202 °C
Density: 1.402 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00007890
CAS Number: 61-78-9
EINECS: 200-518-9
Merck Index: 13,00441

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR