IPTG (isopropyl-β-D-thiogalactopyranoside) for molecular biology dioxane-free
Supplier: PanReac AppliChem
Synonyms: IPTG, (2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol, Isopropyl Beta-D-1-thiogalactopyranoside
A4773.0005 A4773.0025
A4773.0005EA 186 EUR
A4773.0005 A4773.0025
IPTG (isopropyl-β-D-thiogalactopyranoside) for molecular biology dioxane-free
IPTG (Isopropyl-β-D-thiogalactopyranoside)
DNases/RNases/Proteases: not detectable
Assay (HPLC): min. 99 %
α20°C/D; 1 %, H2O: -28.5° - -34.5°
Dioxane: not detectable
Water (K.F.): max. 1.0 %
Formula: C₉H₁₈O₅S
MW: 238.31 g/mol
Boiling Pt: 402
Melting Pt: 109.5…110.5 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063273
CAS Number: 367-93-1
EINECS: 206-703-0