1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,1,1,3,3,3-Hexafluoro-2-methylpropan-2-ol, Hexafluoro-tert-butanol, Hexafluoro-2-methylisopropanol
M-Clarity™ quality level = MQ100
444014-10G 444014-5G
SIAL444014-10GEA 117 EUR
SIAL444014-10G SIAL444014-5G
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol, Sigma-Aldrich®
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol
Formula: C₄H₄F₆O
MW: 182.07 g/mol
Boiling Pt: 60.5…61.5 °C (1013 hPa)
Density: 1.484 g/cm³ (20 °C)
MDL Number: MFCD00039259
CAS Number: 1515-14-6
EINECS: 216-155-4