Coomassie brilliant blue G-250, Proteomics Grade
Supplier: VWR Chemicals
Warning
Synonyms: Brilliant blue G-250, Serva Blue G, Coomassie Blue G 250, Xylene Brilliant Cyanine G, Brillant blue G, Hydrogen [4-[4-(p-ethoxyanilino)-4'-[ethyl(m-sulphonatobenzyl)amino]-2'-methylbenzhydrylene]-3-methylcyclohexa-2,5-dien-1-ylidene](ethyl)(m-sulphonatobenzyl)ammonium monosodium salt, C.I. 42655, Acid Blue 90, Sodium 3-(((4-((4-((4-ethoxyphenyl)amino)phenyl)(4-(ethyl(3-sulphonatobenzyl)iminio)-2-methylcyclohexa-2,5-dien-1-ylidene)methyl)-3-methylphenyl)(ethyl)amino)methyl)benzenesulphonate, Brilliant indocyanine G, Brilliant Blue G
M140-10GEA 78 EUR
M140-10G M140-25G
Coomassie brilliant blue G-250, Proteomics Grade
Coomassie brilliant blue G-250
Formula: C₄₇H₄₈N₃NaO₇S₂
MW: 854.04 g/mol
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00078482
CAS Number: 6104-58-1
EINECS: 228-058-4
Order Now
Specification Test Results
Em (595 nm, Water) >36300
Protease none detected
Protein Staining Pass
Appearance Blue to red crystalline powder