Conductivity standard solutions, Orion™
Supplier: Thermo Fisher Scientific
ORIO011005EA 140 EUR
ORIO011005 663-0024 663-0023 663-0044
Conductivity standard solutions, Orion™
Standards Conductivity Standards
Conductivity standard temperature tables are programmed into Thermo Scientific™ Orion™ meter software for use with the automatic calibration feature.
  • Traceable to NIST standard reference materials
  • Designed to meet all end user requirements, including laboratory grade and DIN standards