Celite® Hyflo Super-cel®, filter aid
Supplier: Thermo Fisher Scientific
Danger
B22658.0E B22658.36 B22658.PG
B22658.0EEA 128 EUR
B22658.0E B22658.36 B22658.PG
Celite® Hyflo Super-cel®, filter aid
Celite® Hyflo® Super Cel, filter aid
CAS No.: 61790-53-2 Odour: Odourless Merck Reference: 14,4973 MDL No.: MFCD00132803
Boiling Pt: 2200 °C (1013 hPa)
Melting Pt: 1710 °C
Density: 2.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00132803
CAS Number: 68855-54-9
EINECS: 319-127-6
Merck Index: 14,4973