Potassium hydrogen phthalate 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1070001
Supplier: VWR Chemicals
87903.290EA 52.3 EUR
87903.290
Potassium hydrogen phthalate 0.2 mol/l in aqueous solution Reag. Ph. Eur. 1070001
Potassium hydrogen phthalate
Ordering information: Supplied with certificate of analysis. Normally manufactured to order. Please check with customer services.
Formula: HOOCC₆H₄COOK
MW: 204,22 g/mol
MDL Number: MFCD00013070
CAS Number: 877-24-7
https://ru.vwr.com/stibo/web/std.lang.all/06/58/16640658.jpg
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022
Order Now
For metal trace analysis in AAS or ICP
Ask for your “customized multi-element standard solution” with your exact composition using the request form
Ask for a quotation
12/31/2022