PRO.TECT Green HD+ Chemical Resistent Gloves, Nitrile
Supplier: UNIGLOVES AERTZE UND KLINIKBED
GA0092 GA0093 GA0094 GA0095
UNIGGA0092EA 30.6 EUR
UNIGGA0092 UNIGGA0093 UNIGGA0094 UNIGGA0095
PRO.TECT Green HD+ Chemical Resistent Gloves, Nitrile
Gloves
Nitrile gloves, latex free, silicone free formulation means no finger marks on metal and glass surfaces.
  • Nitrile formulation conforms to the hand during use for a more comfortable fit
  • Resistant to a range of chemicals
  • Meets medical standards including pinholes
  • Weight: 7,3 g
Extended cuff helps prevent liquids getting into the glove when working in wet conditions and provides additional protection from chemical splashes.
Ambidextrous fit don't have to split a pair to replace a damaged glove.
Beaded cuff provides extra strength during donning and helps prevent liquid roll back. Textured palms and fingers provides improved grip, particularly when working in wet or oily conditions.
Ultra-heavyweight construction is very tough, durable and long lasting. Suitable for use with mobile and tablet displays without removing gloves.
Used in automotive, automotive maintenance and repair, chemical manufacture and use, construction, electronics, engineering, floristry and gardening, general assembly, glass, inspection, laboratory work , light duty maintenance, maintenance, manufacturing and assembly, painting and decorating, pharmaceutical work, repair, utility.
Certifications: Tested to EN ISO 374-1, AQL 1.5 and EN455 parts 1-4, EN ISO 374-5: Virus, UK CA, Food Safe.
Packaging: Each dispenser box contains 100 gloves and each case contains 10 dispenser boxes.