VWR® Flask Brushes
Supplier: VWR Collection
17145-050 17145-052
VWRU17145-050EA 24.4 EUR
VWRU17145-050 VWRU17145-052
VWR® Flask Brushes
Brushes
These brushes with special construction puts the cleaning action directly where it is needed.
The bristles fold flat to fit throught the narrow bottle neck, then expand to fit the chamber.