Titan yellow, Sigma-Aldrich®
Supplier: Merck
Synonyms: Disodium 2,2'-(1-triazene-1,3-diyldi-p-phenylene)bis[6-methylbenzothiazole-7-sulphonate], Direct Yellow 9, Clayton Yellow, Thiazole Yellow G
M-Clarity™ quality level = MQ200
88390-25G 88390-5G
SIAL88390-25GEA 45.6 EUR
SIAL88390-25G SIAL88390-5G
Titan yellow, Sigma-Aldrich®
Titan yellow
Formula: C₂₈H₁₉N₅Na₂O₆S₄
Storage Temperature: Ambient
MDL Number: MFCD00013091
CAS Number: 1829-00-1
EINECS: 217-377-4
Merck Index: 12,09445