Methyl cellulose, 400 cP (C=2%, H₂O)
Supplier: Thermo Fisher Scientific
43147.22
43147.A4
43147.36
43147.22EA
55.5
EUR
43147.22
43147.A4
43147.36
Methyl cellulose, 400 cP (C=2%, H₂O)
Methyl cellulose
CAS No.: 9004-67-5
Physical Form: Powder
Odour: Odourless
Merck Reference: 14,6040
MDL No.: MFCD00081763
Storage Temperature:
Ambient |
MDL Number:
MFCD00081763 CAS Number: 9004-67-5 Merck Index: 14,6040 |