Triethyl orthoacetate, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1,1,1-Triethoxyethane
M-Clarity™ quality level = MQ100
T60402-100ML T60402-2L T60402-500ML
SIALT60402-100MLEA 37.7 EUR
SIALT60402-100ML SIALT60402-2L SIALT60402-500ML
Triethyl orthoacetate, Sigma-Aldrich®
Triethyl orthoacetate
Formula: C₈H₁₈O₃
MW: 162.23 g/mol
Boiling Pt: 142
Density: 0.8847 g/cm³ (20 °C)
Flash Pt: 32
MDL Number: MFCD00009223
CAS Number: 78-39-7
EINECS: 201-112-4