Phthalide, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Isobenzofuran-1(3H)-one
M-Clarity™ quality level = MQ100
P39605-100G P39605-500G
SIALP39605-100GEA 45.3 EUR
SIALP39605-100G SIALP39605-500G
Phthalide, Sigma-Aldrich®
Phthalide
Formula: C₈H₆O₂
MW: 134.13 g/mol
Boiling Pt: 290
Melting Pt: 73
Density: 1.1 g/cm³ (30 °C)
MDL Number: MFCD00005906
CAS Number: 87-41-2
EINECS: 201-744-0