4-Hydroxyphthalic acid ≥98%
Supplier: Thermo Fisher Scientific
Warning
A15091.06 A15091.14 A15091.22
A15091.06EA 121 EUR
A15091.06 A15091.14 A15091.22
4-Hydroxyphthalic acid ≥98%
4-Hydroxyphthalic acid
Purity: 98% CAS No.: 610-35-5 Molecular Formula: C8H6O5 Formula Weight: 182.13 Melting Point: ca 190° dec. MDL No.: MFCD00013984
Formula: C₈H₆O₅
Boiling Pt: 386 °C (1013 hPa)
Melting Pt: ∼ 190 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013984
CAS Number: 610-35-5
EINECS: 210-221-6