Carboxymethyl cellulose sodium salt, low viscosity, Sigma-Aldrich®
Supplier: Merck
Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC
M-Clarity™ quality level = MQ200
C9481-1KG C9481-500G
SIALC9481-1KGEA 564 EUR
SIALC9481-1KG SIALC9481-500G
Carboxymethyl cellulose sodium salt, low viscosity, Sigma-Aldrich®
Carboxymethyl cellulose sodium salt
Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4
Merck Index: 12,01877