Carboxymethyl cellulose sodium salt, medium viscosity, Sigma-Aldrich®
Supplier: Merck
Synonyms: Carboxymethyl cellulose, sodium salt, Cellulose gum, Cellulose, carboxymethyl ether, sodium salt, Sodium carboxymethyl cellulose (Viscosity 800mpa.s), Sodium carboxymethyl cellulose, CMC
M-Clarity™ quality level = MQ100
21902-1KG 21902-250G
SIAL21902-1KGEA 186 EUR
SIAL21902-1KG SIAL21902-250G
Carboxymethyl cellulose sodium salt, medium viscosity, Sigma-Aldrich®
Carboxymethyl cellulose sodium salt
Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
Melting Pt: 300 °C
Storage Temperature: Ambient
MDL Number: MFCD00081472
CAS Number: 9004-32-4
Merck Index: 12,01877