Copper(II) hydroxide 94% stabilised, tech.
Supplier: Thermo Fisher Scientific
Danger
Synonyms: Cupric hydroxide, Cuprichydroxide, Copper dihydroxide
32733.36 32733.A3
32733.36EA 69.7 EUR
32733.36 32733.A3
Copper(II) hydroxide 94% stabilised, tech.
Copper (II) hydroxide
Purity: 94% CAS No.: 20427-59-2 Molecular Formula: H2CuO2 Formula Weight: 97.55 Physical Form: Powder Density: 3.4 Appearance: Blue Odour: Odourless Solubility: Slightly soluble in water Merck Reference: 14,2642 MDL No.: MFCD00010968
Used as a raw material in chemical processing and catalyst manufacturing.
Formula: H₂CuO₂
MW: 97.56
Density: 3.4
Storage Temperature: Ambient
MDL Number: MFCD00010968
CAS Number: 20427-59-2
EINECS: 243-815-9
UN: 3288
ADR: 6.1 II
Merck Index: 14,2642